D(-)-Luciferin Kaliumsalz, Sigma-Aldrich®

Lieferant: Merck

SIAL50227-1MGEA 204 EUR
SIAL50227-1MG
D(-)-Luciferin Kaliumsalz, Sigma-Aldrich®
D(-)-Luciferin Kaliumsalz
Formel: C₁₁H₇KN₂O₃S₂
Molecular Weight: 318,41 g/mol
MDL: MFCD00044928
CAS-Nummer: 115144-35-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR