Ionomycin Calciumsalz

Lieferant: Molekula

39704482-5MG 39704482-1MG
MOLE39704482-5MGEA 0 EUR
MOLE39704482-5MG MOLE39704482-1MG
Ionomycin Calciumsalz
Ionomycin calcium salt
Formel: C₄₁H₇₀CaO₉
MDL: MFCD00151183
CAS-Nummer: 56092-82-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR