1-(2-Tetrahydropyranyl)-3-(trifluormethyl)-1H-pyrazol-5-boronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-6564-250MEA 251 EUR
COBBBB-6564-250M
1-(2-Tetrahydropyranyl)-3-(trifluormethyl)-1H-pyrazol-5-boronsäure
1-(2-Tetrahydropyranyl)-3-(trifluormethyl)-1H-pyrazol-5-boronsäure
Formel: C₉H₁₂BF₃N₂O₃
Molecular Weight: 264,01 g/mol
MDL: MFCD17214248
CAS-Nummer: 1141878-45-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR