2-Aminophenyl-2-pyridylketon

Lieferant: MENAI (PART OF CHEMPUR)

MENASR472.5EA 0 EUR
MENASR472.5 MENASR472.1 MENASR472.2
2-Aminophenyl-2-pyridylketon
2-Aminophenyl-2-pyridylketon
Formel: C₁₂H₁₀N₂O
MDL: MFCD03839744
CAS-Nummer: 42471-56-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR