3-Brombenzaldehydethylenacetal, Sigma-Aldrich®

Lieferant: Merck

SIAL197610-25GEA 63.8 EUR
SIAL197610-25G
3-Brombenzaldehydethylenacetal, Sigma-Aldrich®
3-Brombenzaldehydethylenacetal
Formel: C₉H₉BrO₂
Molecular Weight: 229,07 g/mol
MDL: MFCD00003209
CAS-Nummer: 17789-14-9
EINECS: 241-766-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR