Eicosyltrichlorsilan

Lieferant: SILANE (PART OF CHEMPUR)

SILNSILS08600.10EA 0 EUR
SILNSILS08600.10 SILNSILS08600.25
Eicosyltrichlorsilan
Eicosyltrichlorosilane
Formel: C₂₀H₄₁Cl₃Si
CAS-Nummer: 18733-57-8
EINECS: 242-545-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR