5-Nitro-m-toluidin

Lieferant: BEPHARM LTD.
Gefahr

BEPHB88107.1GEA 0 EUR
BEPHB88107.1G
5-Nitro-m-toluidin
5-Nitro-m-toluidin
Formel: C₇H₈N₂O₂
Molecular Weight: 152,1500 g/mol
MDL: MFCD00082655
CAS-Nummer: 618-61-1
UN: 2660
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR