3-Ethinyltoluol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL521140-1GEA 32.3 EUR
SIAL521140-1G SIAL521140-5G
3-Ethinyltoluol, Sigma-Aldrich®
3-Ethinyltoluol
Formel: C₉H₈
Molecular Weight: 116,16 g/mol
MDL: MFCD01318174
CAS-Nummer: 766-82-5
UN: 3295
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR