2-Hydroxy-4-methoxy-4'-methylbenzophenon

Lieferant: LGC Standards PROMOCHEM

PROOBP251EA 247 EUR
PROOBP251
2-Hydroxy-4-methoxy-4'-methylbenzophenon
2-Hydroxy-4-methoxy-4'-methylbenzophenon
Formel: C₁₅H₁₄O₃
Molecular Weight: 242,2699 g/mol
MDL: MFCD00017172
CAS-Nummer: 1641-17-4
EINECS: 216-688-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR