Phenethyltrichlorsilan

Lieferant: ABCR GMBH

ABCRAB111301.0025EA 0 EUR
ABCRAB111301.0025
Phenethyltrichlorsilan
Phenethyltrichlorsilan
Formel: C₈H₉Cl₃Si
MDL: MFCD00039290
CAS-Nummer: 940-41-0
EINECS: 213-371-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR