Phenethyltrichlorsilan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL420034-1GEA 47.5 EUR
SIAL420034-1G
Phenethyltrichlorsilan, Sigma-Aldrich®
Phenethyltrichlorsilan
Formel: C₈H₉Cl₃Si
MDL: MFCD00039290
CAS-Nummer: 940-41-0
EINECS: 213-371-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR