2-Chlor-p-anisidin

Lieferant: SIA ENAMINE

ENAMEN300-59016.1GEA 0 EUR
ENAMEN300-59016.1G
2-Chlor-p-anisidin
2-Chlor-p-anisidin
Formel: C₇H₈ClNO
Molecular Weight: 157,6000 g/mol
MDL: MFCD00209639
CAS-Nummer: 29242-84-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR