3-(Methylthio)propionsäure

Lieferant: CHEMPUR

CHMPFL13666.POREA 0 EUR
CHMPFL13666.POR
3-(Methylthio)propionsäure
3-(Methylthio)propionsäure
Formel: C₄H₈O₂S
Molecular Weight: 120,17 g/mol
MDL: MFCD00059635
CAS-Nummer: 646-01-5
EINECS: 211-460-9
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR