Dodecacarbonyltriruthenium, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

SIAL245011-5GEA 448 EUR
SIAL245011-5G SIAL245011-1G
Dodecacarbonyltriruthenium, Sigma-Aldrich®
Dodecacarbonyltriruthenium
Formel: C₁₂O₁₂Ru₃
Molecular Weight: 639,33 g/mol
MDL: MFCD00011209
CAS-Nummer: 15243-33-1
EINECS: 239-287-4
UN: 3281
ADR: 6.1,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR