Ethylisocyanatoacetat

Lieferant: FLUOROCHEM LTD (CHEMPUR)

FLUO018685-5GEA 0 EUR
FLUO018685-5G
Ethylisocyanatoacetat
Ethylisocyanatoacetat
Formel: C₅H₇NO₃
Molecular Weight: 129,12 g/mol
MDL: MFCD00002040
CAS-Nummer: 2949-22-6
EINECS: 220-967-4
UN: 2206
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR