Ethylisocyanatoacetat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ200
SIAL238627-10GEA
121
EUR
SIAL238627-10G
SIAL238627-5G
SIAL238627-50G
Ethylisocyanatoacetat, Sigma-Aldrich®
Ethylisocyanatoacetat
Formel:
C₅H₇NO₃ Molecular Weight: 129,12 g/mol |
MDL:
MFCD00002040 CAS-Nummer: 2949-22-6 EINECS: 220-967-4 UN: 2206 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...