5-tert-Butyl-2-hydroxyisophthalaldehyd

Lieferant: Thermo Scientific

MAYBSEW06066.1EA 0 EUR
MAYBSEW06066.1 CHMPSEW06066.250
5-tert-Butyl-2-hydroxyisophthalaldehyd
5-tert-Butyl-2-hydroxyisophthalaldehyd
Formel: C₁₂H₁₄O₃
Molecular Weight: 206,2400 g/mol
MDL: MFCD01567135
CAS-Nummer: 84501-28-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR