5-tert-Butyl-2-hydroxyisophthalaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL554154-5GEA 383 EUR
SIAL554154-5G SIAL554154-1G
5-tert-Butyl-2-hydroxyisophthalaldehyd, Sigma-Aldrich®
5-tert-Butyl-2-hydroxyisophthalaldehyd
Formel: C₁₂H₁₄O₃
Molecular Weight: 206,2400 g/mol
MDL: MFCD01567135
CAS-Nummer: 84501-28-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR