L(-)-Bornylacetat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIALB55203-5GEA
53.8
EUR
SIALB55203-5G
SIALB55203-100G
L(-)-Bornylacetat, Sigma-Aldrich®
L(-)-Bornylacetat
Formel:
C₁₂H₂₀O₂ Molecular Weight: 196,29 g/mol |
MDL:
MFCD00867808 CAS-Nummer: 5655-61-8 EINECS: 227-101-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...