(1-Ethoxycarbonylethyliden)triphenylphosphoran, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL377708-25GEA
150
EUR
SIAL377708-25G
SIAL377708-5G
(1-Ethoxycarbonylethyliden)triphenylphosphoran, Sigma-Aldrich®
(1-Ethoxycarbonylethyliden)triphenylphosphoran
Formel:
C₂₃H₂₃O₂P Molecular Weight: 362,41 g/mol |
MDL:
MFCD00009160 CAS-Nummer: 5717-37-3 EINECS: 227-215-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...