4-Chlorsalicylaldehyd

Lieferant: BEPHARM LTD.

BEPHB4388EA 0 EUR
BEPHB4388
4-Chlorsalicylaldehyd
4-Chlorsalicylaldehyd
Formel: C₇H₅ClO₂
Molecular Weight: 156,5664 g/mol
MDL: MFCD06252499
CAS-Nummer: 2420-26-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR