Koffein, Supelco®
Lieferant: Merck
|
Achtung
|
Synonyme:
Methyltheobromin
, 7-Methyltheobromin
, 1,3,7-Trimethyl-2,6(1H,3H)-purindion
, Coffein
, Thein
, 2,6-Dihydroxy-1,3,7-trimethylpurin
, 3,7-Dihydro-1,3,7-trimethyl-1H-purin-2,6-dion
, 1,3,7-Trimethyl-3,7-dihydro-2H-purin-2,6-dion
, 1,3,7-Trimethylxanthin
, Teein
, Guaranin
, 1,3,7-Trimethyl-1H,3H-purin-2,6-dion
, 1,3,7-Trimethyl-3,7-dihydro-1H-purin-2,6-dion
PHR1095-1GEA
66
EUR
PHR1095-1G
Koffein, Supelco®
Koffein
Formel:
C₈H₁₀N₄O₂ Molecular Weight: 194,19 g/mol Schmelzpunkt: 235…239 °C Dichte: 1,23 g/cm³ (19 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005758 CAS-Nummer: 58-08-2 EINECS: 200-362-1 Merck Index: 13,01636 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...