6-Brom-1-hexanol

Lieferant: MARKET SOURCE PART PROCESS
Achtung

Synonyme: 1-Brom-6-hydroxyhexanHexamethylenbromhydrin

MSPPST01360-1KGEA 0 EUR
MSPPST01360-1KG
6-Brom-1-hexanol
6-Brom-1-hexanol
Formel: BrH₂C(CH₂)₄CH₂OH
Molecular Weight: 181,07 g/mol
Siedepunkt: 105 °C (1013 hPa)
Dichte: 1,3838 g/cm³ (24 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD00002983
CAS-Nummer: 4286-55-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR