Pfefferminzöl, Sigma-Aldrich®

Lieferant: Merck

SIAL77411-25MLEA 37.7 EUR
SIAL77411-25ML
Pfefferminzöl, Sigma-Aldrich®
Peppermint oil
MDL: MFCD00147870
CAS-Nummer: 8006-90-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR