Sonnenblumenkernöl aus Helianthus annuus zur Analyse, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

47123.EA 83.8 EUR
47123.
Sonnenblumenkernöl aus Helianthus annuus zur Analyse, Supelco®
Sunflower seed oil

Characterized sunflower seed oil samples are available for use as controls or check samples for fatty acid methyl ester (FAME) analyses.


  • Neat oil


These reference materials can be used as qualitative standards. They provide an excellent means of standardizing the lipid procedures and comparing the results to others. Packed in an amber ampule under nitrogen. A Certificate of Composition, which includes the chromatographic fingerprint analysis, is provided with the oil sample.

Flammpunkt: >110 °C
MDL: MFCD00132403
CAS-Nummer: 8001-21-6
EINECS: 232-273-9

Bestellen

Spezifikation Testergebnisse

Appearance Clear yellow liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR