Methylrot-Lösung Reag. Ph. Eur. 1055101
Lieferant: VWR Chemicals
Synonyme:
2-(4-(Dimethylamino)phenylazo)benzoesäure
87703.180EA
51.5
EUR
VWRC87703.180_DP
VWRC87703.180_T
87703.180DP
87703.180
Methylrot-Lösung Reag. Ph. Eur. 1055101
Methylrot
Formel:
C₁₅H₁₅N₃O₂ Molecular Weight: 269,3 g/mol |
MDL:
MFCD00002425 CAS-Nummer: 493-52-7 |