Okadasäure Kaliumsalz, Millipore®

Lieferant: Merck Millipore Calbiochem

459618-50EA 187 EUR
459618-50
Okadasäure Kaliumsalz, Millipore®
Okadaic acid potassium salt
Formel: C₄₄H₆₇O₁₃·K
Molecular Weight: 843,11 g/mol
MDL: MFCD00214356
CAS-Nummer: 155751-72-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR