Okadasäure Natriumsalz

Lieferant: UPSTATE TECHNOLOGY

UPST19-130EA 0 EUR
UPST19-130
Okadasäure Natriumsalz
Okadaic acid sodium salt
Formel: C₄₄H₆₇NaO₁₃
Molecular Weight: 827,00 g/mol
MDL: MFCD00210208
CAS-Nummer: 209266-80-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR