3,5-Dimethyl-4-hydroxybenzonitril 97%

Lieferant: Thermo Scientific
Achtung

ACRO353660050EA 109 EUR
ACRO353660050
3,5-Dimethyl-4-hydroxybenzonitril 97%
3,5-Dimethyl-4-hydroxybenzonitril
Formel: C₉H₉NO
Molecular Weight: 147,18 g/mol
Schmelzpunkt: 123…126 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00221716
CAS-Nummer: 4198-90-7

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 123 to 126 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR