3-(Cyclopropylamino)propionitril

Lieferant: FLUOROCHEM LTD (CHEMPUR)

FLUO018484-1GEA 117 EUR
FLUO018484-1G
3-(Cyclopropylamino)propionitril
3-(Cyclopropylamino)propionitril
Formel: C₆H₁₀N₂
MDL: MFCD00060285
CAS-Nummer: 58196-47-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR