3-(Cyclopropylamino)propionitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL560200-1GEA 133 EUR
SIAL560200-1G
3-(Cyclopropylamino)propionitril, Sigma-Aldrich®
3-(Cyclopropylamino)propionitril
Formel: C₆H₁₀N₂
MDL: MFCD00060285
CAS-Nummer: 58196-47-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR