Chlorwasserstoff 1.25 mol/l, Supelco®
Lieferant: Merck
Synonyme:
Chlorwasserstoff
M-Clarity™ Qualitätsstufe = MQ100
17933-250MLEA
114
EUR
17933-250ML
Chlorwasserstoff 1.25 mol/l, Supelco®
Chlorwasserstoff
Formel:
HCl Molecular Weight: 36,46 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011324 CAS-Nummer: 7647-01-0 EINECS: 231-595-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...