Diisobutylaluminiumchlorid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL256803-100GEA
197
EUR
SIAL256803-100G
Diisobutylaluminiumchlorid, Sigma-Aldrich®
Diisobutylaluminiumchlorid
Formel:
C₈H₁₈AlCl Molecular Weight: 176,663 g/mol |
MDL:
MFCD00008927 CAS-Nummer: 1779-25-5 EINECS: 217-216-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...