3-Isoxazolcarbonsäure

Lieferant: SIA ENAMINE

ENAMEN300-22245.1GEA 0 EUR
ENAMEN300-22245.1G
3-Isoxazolcarbonsäure
3-Isoxazolecarboxylic acid
Formel: C₄H₃NO₃
Molecular Weight: 113,07 g/mol
MDL: MFCD06738796
CAS-Nummer: 3209-71-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR