Methyl-2-aminoisonicotinat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL704482-5GEA 119 EUR
SIAL704482-5G SIAL704482-1G
Methyl-2-aminoisonicotinat, Sigma-Aldrich®
Methyl-2-aminoisonicotinat
Formel: C₇H₈N₂O₂
Molecular Weight: 152,15 g/mol
MDL: MFCD04039316
CAS-Nummer: 6937-03-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR