2,2'-Dibromacetophenon

Lieferant: SIA ENAMINE

ENAMEN30001887.1GEA 0 EUR
ENAMEN30001887.5G ENAMEN30001887.10G ENAMEN30001887.1G
2,2'-Dibromacetophenon
2,2'-Dibromacetophenon
Formel: C₈H₆Br₂O
Molecular Weight: 277,9406 g/mol
MDL: MFCD00044660
CAS-Nummer: 49851-55-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR