8-Brom-2-chlorchinolin

Lieferant: SIA ENAMINE

ENAMEN300104450.1GEA 0 EUR
ENAMEN300104450 ENAMEN300104450.1G ENAMEN300104450.5G
8-Brom-2-chlorchinolin
8-Bromo-2-chloroquinoline
Formel: C₉H₅BrClN
Molecular Weight: 242,5000 g/mol
MDL: MFCD08741410
CAS-Nummer: 163485-86-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR