3-Benzylidenphthalid

Lieferant: Molekula

MOLE90029966-500GEA 3770 EUR
MOLE90029966-500G
3-Benzylidenphthalid
3-Benzylidenphthalid
Formel: C₁₅H₁₀O₂
MDL: MFCD00005929
CAS-Nummer: 575-61-1
EINECS: 209-388-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR