3-Benzylidenphthalid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIALB1806-100GEA 82.5 EUR
SIALB1806-100G
3-Benzylidenphthalid, Sigma-Aldrich®
3-Benzylidenphthalid
Formel: C₁₅H₁₀O₂
MDL: MFCD00005929
CAS-Nummer: 575-61-1
EINECS: 209-388-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR