Methyl-3-fluorpicolinat

Lieferant: SIA ENAMINE

ENAMEN300107521EA 0 EUR
ENAMEN300107521 ENAMEN300107521.5G ENAMEN300107521.1G
Methyl-3-fluorpicolinat
Methyl-3-fluorpicolinat
Formel: C₇H₆FNO₂
Molecular Weight: 155,1300 g/mol
MDL: MFCD06659496
CAS-Nummer: 869108-35-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR