2-Brom-4-fluorbenzaldehyd

Lieferant: SIA ENAMINE

ENAMEN300102567.1GEA 0 EUR
ENAMEN300102567 ENAMEN300102567.1G ENAMEN300102567.5G
2-Brom-4-fluorbenzaldehyd
2-Bromo-4-fluorobenzaldehyde
Formel: C₇H₄BrFO
MDL: MFCD00672923
CAS-Nummer: 59142-68-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR