4-Fluor-2-nitrobenzoesäure

Lieferant: SIA ENAMINE

ENAMEN300104066.1GEA 0 EUR
ENAMEN300104066.5G ENAMEN300104066.1G ENAMEN300104066
4-Fluor-2-nitrobenzoesäure
4-Fluoro-2-nitrobenzoic acid
Formel: C₇H₄FNO₄
Molecular Weight: 185,11 g/mol
MDL: MFCD00024300
CAS-Nummer: 394-01-4
EINECS: 206-890-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR