(Tributylstannylmethyl)trimethylsilan

Lieferant: SILANE (PART OF CHEMPUR)
Gefahr

SILNSILS16777.1EA 0 EUR
SILNSILS16777.1 SILNSILS16777.5
(Tributylstannylmethyl)trimethylsilan
(Tributylstannylmethyl)trimethylsilan
Formel: C₁₆H₃₈SiSn
MDL: MFCD00015689
CAS-Nummer: 77425-85-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR