N,N-Diisopropyl-3-methoxyanilin

Lieferant: COMBI-BLOCKS

OT-0762-1G OT-0762-5G
COBBOT-0762-1GEA 204 EUR
COBBOT-0762-1G COBBOT-0762-5G
N,N-Diisopropyl-3-methoxyanilin
N,N-Diisopropyl-3-methoxyanilin
Formel: C₁₃H₂₁NO
MDL: MFCD11504953
CAS-Nummer: 7000-87-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR