N-Fmoc-D-valin

Lieferant: NOVABIOCHEM (PART OF MB)

04-13-1039-25EA 296 EUR
04-13-1039-25 04-13-1039-5
N-Fmoc-D-valin
N-Fmoc-D-valin
Formel: C₂₀H₂₁NO₄
Molecular Weight: 339,39 g/mol
MDL: MFCD00062953
CAS-Nummer: 84624-17-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR