N-Fmoc-L-valin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Fmoc-Val-OH
, N-α-Fmoc-L-Valine
, N-(9-Fluorenylmethoxycarbonyl)-L-valine
SIAL47638-50G-FEA
71.5
EUR
SIAL47638-50G-F
N-Fmoc-L-valin, Sigma-Aldrich®
N-Fmoc-L-valin
Formel:
C₂₀H₂₁NO₄ Molecular Weight: 339,4 g/mol |
MDL:
MFCD00037124 CAS-Nummer: 68858-20-8 EINECS: 272-515-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...