2-Hydroxydimetridazol

Lieferant: EHRENSTORFER GMBH

EHERC12772050EA 570 EUR
EHERC12772050
2-Hydroxydimetridazol
2-Hydroxydimetridazol
Formel: C₅H₇N₃O₃
Molecular Weight: 157,1274 g/mol
MDL: MFCD00159675
CAS-Nummer: 936-05-0
EINECS: 213-312-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR