Ethyl-4-chlor-2-(methylthio)pyrimidin-5-carboxylat

Lieferant: COMBI-BLOCKS INC.

COBBOR-1285-100GEA 177 EUR
COBBOR-1285-100G
Ethyl-4-chlor-2-(methylthio)pyrimidin-5-carboxylat
Ethyl-4-chlor-2-(methylthio)pyrimidin-5-carboxylat
Formel: C₈H₉ClN₂O₂S
Molecular Weight: 232,69 g/mol
MDL: MFCD00006085
CAS-Nummer: 5909-24-0
EINECS: 227-619-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR