N-Boc-L-valin

Lieferant: Molekula

MOLEM51907913EA 0 EUR
MOLEM51907913
N-Boc-L-valin
N-Boc-L-valin
Formel: C₁₀H₁₉NO₄
Molecular Weight: 217,3 g/mol
MDL: MFCD00065605
CAS-Nummer: 13734-41-3
EINECS: 237-307-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR