N-Boc-L-valin
Lieferant: NOVABIOCHEM
Synonyme:
Boc-Val-OH, BOC-L-Valine
8.53011.0100
8.53011.0025
04-12-0029-100EA
95.7
EUR
04-12-0029-100
04-12-0029-25
N-Boc-L-valin
N-Boc-L-valin
Formel:
C₁₀H₁₉NO₄ Molecular Weight: 217,3 g/mol |
MDL:
MFCD00065605 CAS-Nummer: 13734-41-3 EINECS: 237-307-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...